Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGLU16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035822.2 | NP_001030899.1 | 10 - 11 | 1187 | 137 | |
NM_001084847.1 | NP_001078316.1 | 10 - 11 | 1342 | 137 | ||
NM_115876.5 | NP_191572.1 | 11 - 12 | 1654 | 137 | ||
GenBank mRNA | AY045953.1 | - | 11 - 12 | 1378 | 137 | |
AY113935.1 | - | 11 - 12 | 1337 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |