Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340059.1 | NP_001327226.1 | 1 - 2 | 247 | 75 | |
NM_115938.6 | NP_191633.3 | 1 - 2 | 404 | 75 | ||
GenBank mRNA | AF486851.1 | - | 1 - 2 | 208 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |