Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP65-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340060.1 | NP_001319808.1 | 4 - 5 | 661 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |