Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128552.4 | NP_180559.1 | 2 - 3 | 643 | 102 | |
NM_179808.2 | NP_850139.1 | 2 - 3 | 490 | 102 | ||
GenBank mRNA | AY063966.1 | - | 2 - 3 | 505 | 102 | |
AY096462.1 | - | 2 - 3 | 399 | 102 | ||
D17579.1 | - | 2 - 3 | 481 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |