Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128578.4 | NP_180584.1 | 2 - 3 | 600 | 79 | |
GenBank mRNA | AF418309.1 | - | 2 - 3 | 351 | 79 | |
AY062720.1 | - | 2 - 3 | 460 | 79 | ||
AY114650.1 | - | 2 - 3 | 351 | 79 | ||
AY136318.1 | - | 2 - 3 | 460 | 79 | ||
BT002338.1 | - | 2 - 3 | 351 | 79 | ||
BT008482.1 | - | 2 - 3 | 351 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |