Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: U2B'' | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336249.1 | NP_001324159.1 | 2 - 3 | 472 | 95 | |
NM_128579.4 | NP_180585.1 | 2 - 3 | 472 | 95 | ||
GenBank mRNA | AK227609.1 | - | 2 - 3 | 316 | 95 | |
BT003068.1 | - | 2 - 3 | 241 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Arabidopsis thaliana Probe spans exons |