Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121309.6 | NP_196810.5 | 5 - 6 | 908 | 92 | |
GenBank mRNA | FJ422582.1 | - | 5 - 6 | 860 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |