Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343293.1 | NP_001330207.1 | 4 - 5 | 570 | 86 | |
NM_121382.3 | NP_196883.1 | 4 - 5 | 467 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |