Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BET9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203379.1 | NP_001190308.1 | 7 - 8 | 2152 | 98 | |
NM_001343329.1 | NP_001332743.1 | 8 - 9 | 1930 | 98 | ||
NM_121431.3 | NP_568297.1 | 7 - 8 | 2154 | 98 | ||
GenBank mRNA | AY059785.1 | - | 7 - 8 | 1998 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |