Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXE16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121435.2 | NP_568298.1 | 1 - 2 | 1228 | 69 | |
GenBank mRNA | AY085477.1 | - | 1 - 2 | 1071 | 69 | |
BT000439.1 | - | 1 - 2 | 1066 | 69 | ||
BT001209.1 | - | 1 - 2 | 799 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |