Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121589.2 | NP_197088.1 | 1 - 2 | 1154 | 70 | |
GenBank mRNA | AY086574.1 | - | 1 - 2 | 827 | 70 | |
BT001926.1 | - | 1 - 2 | 766 | 70 | ||
X94937.1 | - | 766 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Inventoried Probe spans exons |