Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLN1;4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343456.1 | NP_001331815.1 | 3 - 4 | 312 | 76 | |
NM_121663.3 | NP_568335.1 | 3 - 4 | 446 | 76 | ||
GenBank mRNA | AY059932.1 | - | 3 - 4 | 263 | 76 | |
AY128749.1 | - | 3 - 4 | 220 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |