Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036817.3 | NP_001031894.1 | 5 - 6 | 877 | 130 | |
NM_001343458.1 | NP_001332636.1 | 5 - 6 | 877 | 130 | ||
NM_121669.3 | NP_197166.2 | 5 - 6 | 752 | 130 | ||
GenBank mRNA | AY062755.1 | - | 5 - 6 | 725 | 130 | |
BT010359.1 | - | 4 - 5 | 582 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |