Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036817.3 | NP_001031894.1 | 8 - 9 | 1694 | 76 | |
NM_001343458.1 | NP_001332636.1 | 8 - 9 | 1694 | 76 | ||
NM_121669.3 | NP_197166.2 | 8 - 9 | 1569 | 76 | ||
GenBank mRNA | AY062755.1 | - | 8 - 9 | 1542 | 76 | |
BT010359.1 | - | 7 - 8 | 1399 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |