Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343464.1 | NP_001318573.1 | 9 - 10 | 1991 | 105 | |
NM_001343465.1 | NP_001330040.1 | 9 - 10 | 1988 | 105 | ||
NM_121684.4 | NP_197180.1 | 9 - 10 | 1988 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |