Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPO1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203395.1 | NP_001190324.1 | 23 - 24 | 2385 | 113 | |
NM_121708.3 | NP_197204.1 | 24 - 25 | 2522 | 113 | ||
GenBank mRNA | AY056126.1 | - | 24 - 25 | 2418 | 113 | |
AY096633.1 | - | 24 - 25 | 2250 | 113 | ||
Y18469.1 | - | 2345 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |