Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARFB1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343479.1 | NP_001331669.1 | 2 - 3 | 464 | 135 | |
NM_121712.4 | NP_197208.1 | 2 - 3 | 464 | 135 | ||
GenBank mRNA | AY093231.1 | - | 2 - 3 | 309 | 135 | |
BT003403.1 | - | 2 - 3 | 149 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |