Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121748.4 | NP_197244.1 | 6 - 7 | 1352 | 71 | |
GenBank mRNA | AY139754.1 | - | 6 - 7 | 1304 | 71 | |
BT004543.1 | - | 6 - 7 | 1259 | 71 | ||
KJ138681.1 | - | 6 - 7 | 1259 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |