Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121816.2 | NP_197312.1 | 2 - 3 | 323 | 132 | |
GenBank mRNA | AY092964.1 | - | 2 - 3 | 323 | 132 | |
AY093208.1 | - | 2 - 3 | 319 | 132 | ||
AY114562.1 | - | 2 - 3 | 230 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |