Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FASS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121863.4 | NP_568364.1 | 4 - 5 | 570 | 92 | |
GenBank mRNA | AF280057.1 | - | 4 - 5 | 546 | 92 | |
AF360316.1 | - | 4 - 5 | 533 | 92 | ||
AY113888.1 | - | 4 - 5 | 388 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |