Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121951.5 | NP_197447.2 | 1 - 2 | 636 | 119 | |
GenBank mRNA | AY074342.1 | - | 1 - 2 | 286 | 119 | |
AY117160.1 | - | 1 - 2 | 273 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |