Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343605.1 | NP_001318599.1 | 2 - 3 | 390 | 153 | |
NM_121952.3 | NP_197448.2 | 2 - 3 | 422 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |