Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPUORF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036377.1 | NP_001031454.1 | 6 - 7 | 1110 | 84 | |
NM_128684.5 | NP_180686.2 | 6 - 7 | 1379 | 84 | ||
GenBank mRNA | AY954808.1 | - | 6 - 7 | 519 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |