Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LBD14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128688.4 | NP_180689.1 | 1 - 2 | 344 | 96 | |
GenBank mRNA | AB473850.1 | - | 1 - 2 | 344 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |