Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLX2-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128692.5 | NP_180693.1 | 6 - 7 | 1111 | 116 | |
NM_179835.1 | NP_850166.1 | 6 - 7 | 885 | 116 | ||
GenBank mRNA | AY072200.1 | - | 6 - 7 | 888 | 116 | |
AY096672.1 | - | 6 - 7 | 730 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |