Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPK24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128707.3 | NP_180708.1 | 6 - 7 | 1764 | 115 | |
GenBank mRNA | DQ446585.1 | - | 6 - 7 | 1486 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |