Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336351.1 | NP_001318329.1 | 21 - 22 | 3019 | 91 | |
NM_001336352.1 | NP_001323600.1 | 19 - 20 | 2306 | 91 | ||
NM_001336353.1 | NP_001323601.1 | 14 - 15 | 1532 | 91 | ||
NM_001336354.1 | NP_001323599.1 | 17 - 18 | 2097 | 91 | ||
NM_128748.3 | NP_180749.2 | 19 - 20 | 2306 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |