Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUR ALPHA-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128768.3 | NP_565736.1 | 4 - 5 | 867 | 114 | |
NM_179851.3 | NP_850182.1 | 4 - 5 | 1119 | 114 | ||
GenBank mRNA | AF136152.1 | - | 4 - 5 | 863 | 114 | |
AF419556.1 | - | 4 - 5 | 847 | 114 | ||
AF446882.1 | - | 4 - 5 | 737 | 114 | ||
AY052714.1 | - | 4 - 5 | 851 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |