Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATZW10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128850.3 | NP_565757.2 | 5 - 6 | 1407 | 65 | |
GenBank mRNA | AY069883.1 | - | 5 - 6 | 1341 | 65 | |
U80984.1 | - | 5 - 6 | 1329 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |