Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336459.1 | NP_001325202.1 | 7 - 8 | 3762 | 82 | |
NM_128914.3 | NP_180912.2 | 8 - 9 | 3637 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |