Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLO5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128925.2 | NP_180923.1 | 5 - 6 | 768 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |