Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MNS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104037.4 | NP_175570.1 | 2 - 3 | 383 | 90 | |
GenBank mRNA | AY081353.1 | - | 2 - 3 | 370 | 90 | |
AY128845.1 | - | 2 - 3 | 197 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |