Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B ALPHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198270.1 | NP_001185199.1 | 9 - 10 | 1165 | 81 | |
NM_001333494.1 | NP_001322512.1 | 7 - 8 | 3064 | 81 | ||
NM_001333495.1 | NP_001322511.1 | 9 - 10 | 1186 | 81 | ||
NM_104047.4 | NP_564595.1 | 8 - 9 | 2793 | 81 | ||
NM_202274.2 | NP_974003.1 | 7 - 8 | 1087 | 81 | ||
GenBank mRNA | AK230310.1 | - | 8 - 9 | 2573 | 81 | |
AY120756.1 | - | 7 - 8 | 1088 | 81 | ||
BT000109.1 | - | 7 - 8 | 846 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |