Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340110.1 | NP_001327121.1 | 1 - 2 | 418 | 84 | |
NM_116022.5 | NP_191716.2 | 1 - 2 | 495 | 84 | ||
GenBank mRNA | AK226722.1 | - | 1 - 2 | 358 | 84 | |
BT002756.1 | - | 1 - 2 | 199 | 84 | ||
EU249332.1 | - | 1 - 2 | 199 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |