Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340110.1 | NP_001327121.1 | 5 - 6 | 865 | 101 | |
NM_116022.5 | NP_191716.2 | 5 - 6 | 942 | 101 | ||
GenBank mRNA | AK226722.1 | - | 5 - 6 | 805 | 101 | |
BT002756.1 | - | 5 - 6 | 646 | 101 | ||
EU249332.1 | - | 5 - 6 | 646 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |