Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DL1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340126.1 | NP_001325984.1 | 7 - 8 | 750 | 103 | |
NM_001340127.1 | NP_001325985.1 | 7 - 8 | 750 | 103 | ||
NM_116041.3 | NP_191735.2 | 7 - 8 | 750 | 103 | ||
GenBank mRNA | AY189279.1 | - | 7 - 8 | 674 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |