Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084860.1 | NP_001078329.1 | 1 - 2 | 651 | 70 | |
NM_001203231.1 | NP_001190160.1 | 2 - 3 | 526 | 70 | ||
NM_001340137.1 | NP_001327149.1 | 1 - 2 | 651 | 70 | ||
NM_116074.2 | NP_191768.2 | 2 - 3 | 526 | 70 | ||
GenBank mRNA | BT033108.1 | - | 1 - 2 | 437 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |