Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340142.1 | NP_001327193.1 | 6 - 7 | 2193 | 69 | |
NM_001340143.1 | NP_001327192.1 | 7 - 8 | 2218 | 69 | ||
NM_116080.3 | NP_191774.2 | 7 - 8 | 2236 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |