Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATGSTF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116141.2 | NP_191835.1 | 1 - 2 | 215 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |