Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSP41A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116179.5 | NP_191873.1 | 1 - 2 | 566 | 78 | |
GenBank mRNA | AF428269.1 | - | 1 - 2 | 425 | 78 | |
AY059887.1 | - | 1 - 2 | 488 | 78 | ||
AY128804.1 | - | 1 - 2 | 397 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |