Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340195.1 | NP_001319825.1 | 3 - 4 | 678 | 68 | |
NM_116190.5 | NP_191884.1 | 3 - 4 | 568 | 68 | ||
NM_202753.2 | NP_974482.1 | 2 - 3 | 964 | 68 | ||
GenBank mRNA | AF428402.1 | - | 3 - 4 | 515 | 68 | |
AK227627.1 | - | 3 - 4 | 517 | 68 | ||
BT010165.1 | - | 3 - 4 | 442 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |