Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNE10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340226.1 | NP_001329601.1 | 1 - 2 | 732 | 95 | |
NM_001340227.1 | NP_001329602.1 | 2 - 3 | 657 | 95 | ||
NM_116222.4 | NP_191916.3 | 2 - 3 | 802 | 95 | ||
GenBank mRNA | AK117229.1 | - | 2 - 3 | 649 | 95 | |
BT025971.1 | - | 2 - 3 | 578 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |