Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YAB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084868.2 | NP_001078337.1 | 4 - 5 | 974 | 73 | |
NM_116235.3 | NP_567154.1 | 5 - 6 | 1058 | 73 | ||
GenBank mRNA | AF136540.1 | - | 5 - 6 | 679 | 73 | |
AK316696.1 | - | 4 - 5 | 569 | 73 | ||
AY037186.1 | - | 5 - 6 | 863 | 73 | ||
BT002662.1 | - | 5 - 6 | 571 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |