Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340260.1 | NP_001328554.1 | 1 - 2 | 1065 | 78 | |
NM_116293.3 | NP_567182.1 | 2 - 3 | 736 | 78 | ||
GenBank mRNA | AY086598.1 | - | 2 - 3 | 363 | 78 | |
BT024819.1 | - | 1 - 2 | 272 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |