Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125435.1 | NP_001118907.1 | 5 - 6 | 1704 | 68 | |
NM_116298.4 | NP_567183.2 | 5 - 6 | 1808 | 68 | ||
GenBank mRNA | AK226968.1 | - | 5 - 6 | 1649 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |