Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APRR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116302.4 | NP_191985.3 | 8 - 9 | 2241 | 128 | |
GenBank mRNA | AK229225.1 | - | 8 - 9 | 2241 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |