Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APRR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340267.1 | NP_001328422.1 | 3 - 4 | 648 | 94 | |
NM_116302.4 | NP_191985.3 | 11 - 12 | 2809 | 94 | ||
GenBank mRNA | AK229225.1 | - | 11 - 12 | 2809 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |