Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SWN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340355.1 | NP_001328437.1 | 11 - 12 | 2001 | 107 | |
NM_001340356.1 | NP_001328436.1 | 11 - 12 | 1998 | 107 | ||
NM_001340357.1 | NP_001328435.1 | 11 - 12 | 1998 | 107 | ||
NM_116433.3 | NP_567221.1 | 11 - 12 | 2018 | 107 | ||
GenBank mRNA | AF100163.1 | - | 11 - 12 | 1972 | 107 | |
AY057477.1 | - | 11 - 12 | 1998 | 107 | ||
AY090293.1 | - | 11 - 12 | 1905 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |