Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340373.1 | NP_001329308.1 | 17 - 18 | 1980 | 87 | |
NM_001340374.1 | NP_001329307.1 | 16 - 17 | 2047 | 87 | ||
NM_116472.4 | NP_192148.2 | 17 - 18 | 2154 | 87 | ||
GenBank mRNA | Z48243.1 | - | 1944 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |