Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203731.2 | NP_001190660.1 | 4 - 5 | 415 | 93 | |
NM_116490.4 | NP_192165.1 | 3 - 4 | 558 | 93 | ||
GenBank mRNA | AK226746.1 | - | 3 - 4 | 468 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |